Assay Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172485.3 | NP_766073.2 | 4 - 5 | 1528 | 82 | |
| GenBank mRNA | AK047508.1 | - | 1314 | 82 | ||
| AK051714.1 | - | 4 - 5 | 1370 | 82 | ||
| AK083798.1 | - | 4 - 5 | 1356 | 82 | ||
| BC063250.1 | - | 4 - 5 | 1358 | 6410178 | 82 | |
More Information
Set Membership: |
|

Thsd7b
Genomic Map