Assay Details
| Gene Symbol: Dcaf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172502.3 | NP_766090.2 | 3 - 4 | 380 | 63 | |
| GenBank mRNA | AK032982.1 | - | 3 - 4 | 392 | 63 | |
| AK144808.1 | - | 3 - 4 | 383 | 63 | ||
| AK153305.1 | - | 3 - 4 | 371 | 63 | ||
| AK155000.1 | - | 3 - 4 | 381 | 63 | ||
| BC057552.1 | - | 3 - 4 | 381 | 6403172 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf15
Genomic Map