Assay Details
| Gene Symbol: Dcaf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172502.3 | NP_766090.2 | 11 - 12 | 1641 | 72 | |
| GenBank mRNA | AK032982.1 | - | 11 - 12 | 1653 | 72 | |
| AK144808.1 | - | 11 - 12 | 1644 | 72 | ||
| AK153305.1 | - | 11 - 12 | 1632 | 72 | ||
| AK155000.1 | - | 11 - 12 | 1642 | 72 | ||
| BC057552.1 | - | 11 - 12 | 1642 | 6403172 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543155 | AK027972.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130594 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120190 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170445 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153916 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018181 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120190 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170445 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153916 | - | - |
| Whole Mouse Genome G4122A | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | BC057552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK144808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK153305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | AK155000.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412543 | NM_172502.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115818 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153916 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170445 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06993577 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06993573 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07364401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06711202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06711199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06793927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06986594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06986584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06793922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06793935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07073400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06793934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018160 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580094 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454750_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580085 | - | - |
| Mouse Genome 430 2.0 Array | 1454750_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1442132_at | - | - |
| Mouse Genome 430 2.0 Array | 1442132_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454750_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212123_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212123_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800019565.mm.1 | NM_172502.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019565.mm.1 | NM_172502.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019565.mm.1 | TC0800002569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019566.mm.1 | NM_172502.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019566.mm.1 | NM_172502.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019566.mm.1 | TC0800002569.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454750_a_at | - | - |
| Mouse Expression Set 430 | 1454750_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454750_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454750_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454750_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454750_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 653000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6650053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706788 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706788 | - | - |
| mouse-6 v1.1 (Illumina) | scl34569.13_162-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34569.13_162-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2896042 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896042 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34569.13_162-S | - | - |

Dcaf15
Genomic Map