Assay Details
| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172503.3 | NP_766091.2 | 1 - 2 | 178 | 83 | |
| GenBank mRNA | AK041934.1 | - | 1 - 2 | 192 | 83 | |
| AK052168.1 | - | 1 - 2 | 192 | 83 | ||
| AK053663.1 | - | 1 - 2 | 192 | 83 | ||
| AK150221.1 | - | 1 - 2 | 192 | 83 | ||
| AK155465.1 | - | 1 - 2 | 192 | 83 | ||
| BC058080.1 | - | 1 - 2 | 178 | 6822376 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim4
Genomic Map