Assay Details
| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172503.3 | NP_766091.2 | 7 - 8 | 1560 | 80 | |
| GenBank mRNA | AK052168.1 | - | 7 - 8 | 1574 | 80 | |
| AK053663.1 | - | 7 - 8 | 1574 | 80 | ||
| AK131119.1 | - | 2 - 3 | 418 | 80 | ||
| AK150221.1 | - | 7 - 8 | 1575 | 80 | ||
| BC058080.1 | - | 7 - 8 | 1560 | 6822376 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim4
Genomic Map