Assay Details
| Gene Symbol: Dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172508.2 | NP_766096.1 | 2 - 3 | 936 | 67 | |
| GenBank mRNA | AK031737.1 | - | 2 - 3 | 623 | 67 | |
| AK045065.1 | - | 2 - 3 | 843 | 67 | ||
| AK153326.1 | - | 2 - 3 | 609 | 67 | ||
| BC053095.1 | - | 2 - 3 | 936 | 6401066 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dse
Genomic Map