Assay Details
| Gene Symbol: Abhd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272070.1 | NP_001258999.1 | 3 - 4 | 473 | 73 | |
| NM_172511.4 | NP_766099.3 | 3 - 4 | 473 | 73 | ||
| NR_073572.1 | 2 - 3 | 289 | 73 | |||
| GenBank mRNA | AK052670.1 | - | 3 - 4 | 422 | 73 | |
| AK084998.1 | - | 3 - 4 | 421 | 73 | ||
| BC058347.1 | - | 3 - 4 | 422 | 6820515 | 73 | |
| CX237340.1 | - | 2 - 3 | 236 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Abhd10
Genomic Map