Assay Details
| Gene Symbol: Arhgef19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172520.2 | NP_766108.1 | 11 - 12 | 1962 | 69 | |
| GenBank mRNA | AK031443.1 | - | 11 - 12 | 1963 | 69 | |
| AK053075.1 | - | 11 - 12 | 1963 | 69 | ||
| BC060376.1 | - | 9 - 10 | 1692 | 5340007 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef19
Genomic Map