Assay Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172523.3 | NP_766111.1 | 3 - 4 | 74 | ||
| GenBank mRNA | AJ555564.1 | - | 2 - 3 | 74 | ||
| AK034368.1 | - | 2 - 3 | 74 | |||
| AK035644.1 | - | 3 - 4 | 74 | |||
| AK041274.1 | - | 3 - 4 | 74 | |||
| BC078449.1 | - | 3 - 4 | 5705966 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a2
Genomic Map