Assay Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172523.3 | NP_766111.1 | 10 - 11 | 1152 | 56 | |
| GenBank mRNA | AJ555564.1 | - | 9 - 10 | 1016 | 56 | |
| AK033871.1 | - | 10 - 11 | 1108 | 56 | ||
| AK034368.1 | - | 5 - 6 | 2511 | 56 | ||
| AK041274.1 | - | 10 - 11 | 1104 | 56 | ||
| BC078449.1 | - | 9 - 10 | 1023 | 5705966 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a2
Genomic Map