Assay Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172523.3 | NP_766111.1 | 12 - 13 | 1281 | 82 | |
| GenBank mRNA | AJ555564.1 | - | 11 - 12 | 1145 | 82 | |
| AK033871.1 | - | 12 - 13 | 1237 | 82 | ||
| AK034368.1 | - | 7 - 8 | 2640 | 82 | ||
| AK035644.1 | - | 12 - 13 | 1265 | 82 | ||
| AK041274.1 | - | 12 - 13 | 1233 | 82 | ||
| BC078449.1 | - | 11 - 12 | 1152 | 5705966 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc18a2
Genomic Map