Assay Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146048.1 | NP_001139520.1 | 7 - 8 | 868 | 66 | |
| NM_172528.3 | NP_766116.3 | 7 - 8 | 516 | 66 | ||
| GenBank mRNA | AK051043.1 | - | 7 - 8 | 867 | 66 | |
| AK082721.1 | - | 7 - 8 | 517 | 66 | ||
| AK147075.1 | - | 7 - 8 | 868 | 66 | ||
| AK220166.1 | - | 7 - 8 | 886 | 66 | ||
| BC046591.1 | - | 7 - 8 | 855 | 66 | ||
| BC087542.1 | - | 7 - 8 | 715 | 4934980 | 66 | |
More Information
Set Membership: |
|

Lrrc1
Genomic Map