Assay Details
| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012518.3 | NP_001012536.2 | 21 - 22 | 3210 | 82 | |
| NM_001109686.2 | NP_001103156.1 | 20 - 21 | 3051 | 82 | ||
| NM_001109687.2 | NP_001103157.1 | 20 - 21 | 3066 | 82 | ||
| NM_172545.4 | NP_766133.2 | 21 - 22 | 3189 | 82 | ||
| GenBank mRNA | AB205007.1 | - | 21 - 22 | 3216 | 82 | |
| AK049454.1 | - | 18 - 19 | 2460 | 82 | ||
| AK052174.1 | - | 21 - 22 | 3173 | 82 | ||
| AK082062.1 | - | 21 - 22 | 3200 | 82 | ||
| AK129461.1 | - | 2 - 3 | 301 | 82 | ||
| AK149125.1 | - | 1 - 2 | 133 | 82 | ||
| BC023904.1 | - | 3 - 4 | 455 | 82 | ||
| BC056938.1 | - | 18 - 19 | 2767 | 82 | ||
| BC089302.1 | - | 18 - 19 | 2919 | 82 | ||
More Information
Set Membership: |
|

Ehmt1
Genomic Map