Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 4 - 5 | 903 | 89 | |
| GenBank mRNA | AK030039.1 | - | 4 - 5 | 907 | 89 | |
| AK042071.1 | - | 2 - 3 | 460 | 89 | ||
| AK047545.1 | - | 4 - 5 | 896 | 89 | ||
| AK053356.1 | - | 4 - 5 | 894 | 89 | ||
| AK085666.1 | - | 4 - 5 | 896 | 89 | ||
| AK128956.1 | - | 4 - 5 | 895 | 89 | ||
| AK154181.1 | - | 4 - 5 | 894 | 89 | ||
| AK154729.1 | - | 4 - 5 | 885 | 89 | ||
| BC110697.1 | - | 4 - 5 | 879 | 30356222 | 89 | |
| BC117939.1 | - | 4 - 5 | 896 | 89 | ||
| BC117940.1 | - | 4 - 5 | 896 | 40097816 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map