Assay Details
| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 11 - 12 | 2704 | 73 | |
| GenBank mRNA | AK003792.2 | - | 2 - 3 | 442 | 73 | |
| AK030039.1 | - | 10 - 11 | 2486 | 73 | ||
| AK042071.1 | - | 9 - 10 | 2261 | 73 | ||
| AK053356.1 | - | 11 - 12 | 2695 | 73 | ||
| AK085666.1 | - | 11 - 12 | 2697 | 73 | ||
| AK154729.1 | - | 11 - 12 | 2686 | 73 | ||
| BC110697.1 | - | 11 - 12 | 2680 | 30356222 | 73 | |
| BC117939.1 | - | 11 - 12 | 2697 | 73 | ||
| BC117940.1 | - | 11 - 12 | 2697 | 40097816 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polrmt
Genomic Map