Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 11 - 12 | 2465 | 56 | |
| GenBank mRNA | AK036710.1 | - | 10 - 11 | 2285 | 56 | |
| AK147774.1 | - | 11 - 12 | 2351 | 57 | ||
| BC058512.1 | - | 11 - 12 | 1298 | 6817014 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntrob
Genomic Map