Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 13 - 14 | 2894 | 65 | |
| GenBank mRNA | AK036710.1 | - | 12 - 13 | 2714 | 65 | |
| AK147774.1 | - | 13 - 14 | 2784 | 65 | ||
| BC058512.1 | - | 13 - 14 | 1727 | 6817014 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894310 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894314 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750326 | AK028850.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750326 | BC025043.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32613 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124706 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119573 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894307 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07385101 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07385086 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119573 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07385058 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124706 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07385112 | - | - |
| Whole Mouse Genome G4122A | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | AK147774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | BC058512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | AK036710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224099 | NM_172560.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108576 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124706 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119573 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894310 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09224179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09224170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09573830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09728298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09563092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09224180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09226922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27894312 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387306 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387299 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387315 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387304 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433958_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387295 | - | - |
| Mouse Genome 430 2.0 Array | 1433958_at | - | - |
| Mouse Genome 430 2.0 Array | 1433958_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216846_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216846_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100027589.mm.1 | NM_172560.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027589.mm.1 | NM_172560.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027589.mm.1 | TC1100003035.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433958_PM_at | - | - |
| Mouse Expression Set 430 | 1433958_at | - | - |
| Murine Genome U74v2 Set | 96230_PM_at | - | - |
| Murine Genome U74v2 Set | 96230_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433958_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433958_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433958_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433958_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 440552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050440 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050440 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700768 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700768 | - | - |
| mouse-6 v1.1 (Illumina) | scl40039.20.1_60-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050440 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40039.20.1_60-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2700768 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2700768 | - | - |

Cntrob
Genomic Map