Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 14 - 15 | 3036 | 110 | |
| GenBank mRNA | AK036710.1 | - | 13 - 14 | 2856 | 110 | |
| AK147774.1 | - | 14 - 15 | 2926 | 110 | ||
| BC058512.1 | - | 14 - 15 | 1869 | 6817014 | 110 | |
More Information
Set Membership: |
|

Cntrob
Genomic Map