Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 15 - 16 | 3098 | 59 | |
| GenBank mRNA | AK034947.1 | - | 1 - 2 | 722 | 59 | |
| AK036710.1 | - | 14 - 15 | 2918 | 59 | ||
| AK147774.1 | - | 15 - 16 | 2988 | 59 | ||
| BC058512.1 | - | 15 - 16 | 1931 | 6817014 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntrob
Genomic Map