Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167680.1 | NP_001161152.1 | 6 - 7 | 977 | 76 | |
| NM_172572.3 | NP_766160.2 | 6 - 7 | 1191 | 76 | ||
| GenBank mRNA | AK028872.1 | - | 6 - 7 | 1192 | 76 | |
| AK042365.1 | - | 5 - 6 | 1644 | 76 | ||
| AK154639.1 | - | 6 - 7 | 957 | 76 | ||
| AK171085.1 | - | 6 - 7 | 973 | 76 | ||
| AY167043.1 | - | 6 - 7 | 977 | 76 | ||
| BC052182.1 | - | 6 - 7 | 925 | 6335022 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map