Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167680.1 | NP_001161152.1 | 9 - 10 | 1430 | 91 | |
| NM_172572.3 | NP_766160.2 | 9 - 10 | 1644 | 91 | ||
| GenBank mRNA | AK028872.1 | - | 9 - 10 | 1645 | 91 | |
| AK154639.1 | - | 9 - 10 | 1410 | 91 | ||
| AK171085.1 | - | 9 - 10 | 1426 | 91 | ||
| AY167043.1 | - | 9 - 10 | 1430 | 91 | ||
| BC031398.1 | - | 3 - 4 | 463 | 91 | ||
| BC052182.1 | - | 9 - 10 | 1378 | 6335022 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map