Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167680.1 | NP_001161152.1 | 12 - 13 | 1774 | 66 | |
| NM_172572.3 | NP_766160.2 | 12 - 13 | 1988 | 66 | ||
| GenBank mRNA | AK028872.1 | - | 12 - 13 | 1989 | 66 | |
| AK154639.1 | - | 12 - 13 | 1754 | 66 | ||
| AK171085.1 | - | 12 - 13 | 1770 | 66 | ||
| AY167043.1 | - | 12 - 13 | 1774 | 66 | ||
| BC031398.1 | - | 6 - 7 | 807 | 66 | ||
| BC052182.1 | - | 12 - 13 | 1722 | 6335022 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map