Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167680.1 | NP_001161152.1 | 13 - 14 | 1877 | 57 | |
| NM_172572.3 | NP_766160.2 | 13 - 14 | 2091 | 57 | ||
| GenBank mRNA | AK028872.1 | - | 13 - 14 | 2092 | 57 | |
| AK154639.1 | - | 13 - 14 | 1857 | 57 | ||
| AK171085.1 | - | 13 - 14 | 1873 | 57 | ||
| AY167043.1 | - | 13 - 14 | 1877 | 57 | ||
| BC031398.1 | - | 7 - 8 | 910 | 57 | ||
| BC052182.1 | - | 13 - 14 | 1825 | 6335022 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map