Assay Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172573.2 | NP_766161.1 | 7 - 8 | 1043 | 61 | |
| GenBank mRNA | AK048667.1 | - | 7 - 8 | 1045 | 61 | |
| AK084254.1 | - | 7 - 8 | 1176 | 61 | ||
| AK167142.1 | - | 7 - 8 | 1015 | 61 | ||
| AK171813.1 | - | 2 - 3 | 367 | 61 | ||
| BC060640.1 | - | 6 - 7 | 867 | 61 | ||
| BC120923.1 | - | 7 - 8 | 1041 | 61 | ||
| BC120924.1 | - | 7 - 8 | 1045 | 40097903 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Engase
Genomic Map