Assay Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172582.3 | NP_766170.2 | 6 - 7 | 824 | 90 | |
| GenBank mRNA | AK077863.1 | - | 6 - 7 | 748 | 90 | |
| AK131889.1 | - | 6 - 7 | 786 | 90 | ||
| AK167380.1 | - | 6 - 7 | 752 | 90 | ||
| AK170311.1 | - | 5 - 6 | 1289 | 90 | ||
| BC024135.1 | - | 6 - 7 | 725 | 90 | ||
| BC051055.1 | - | 6 - 7 | 740 | 90 | ||
| BC057914.1 | - | 3 - 4 | 488 | 90 | ||
| BC106183.1 | - | 6 - 7 | 761 | 90 | ||
| BC132168.1 | - | 6 - 7 | 824 | 40130445 | 90 | |
| BC132170.1 | - | 6 - 7 | 824 | 40130447 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq6
Genomic Map