Assay Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172582.3 | NP_766170.2 | 7 - 8 | 893 | 57 | |
| GenBank mRNA | AK077863.1 | - | 7 - 8 | 817 | 57 | |
| AK131889.1 | - | 7 - 8 | 855 | 57 | ||
| AK167380.1 | - | 7 - 8 | 821 | 57 | ||
| AK170311.1 | - | 6 - 7 | 1358 | 57 | ||
| BC024135.1 | - | 7 - 8 | 794 | 57 | ||
| BC051055.1 | - | 7 - 8 | 809 | 57 | ||
| BC057914.1 | - | 4 - 5 | 557 | 57 | ||
| BC132168.1 | - | 7 - 8 | 893 | 40130445 | 57 | |
| BC132170.1 | - | 7 - 8 | 893 | 40130447 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq6
Genomic Map