Assay Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172582.3 | NP_766170.2 | 9 - 10 | 1203 | 76 | |
| GenBank mRNA | AK077863.1 | - | 9 - 10 | 1127 | 76 | |
| AK131889.1 | - | 9 - 10 | 1165 | 76 | ||
| AK167380.1 | - | 9 - 10 | 1131 | 76 | ||
| AK170311.1 | - | 8 - 9 | 1668 | 76 | ||
| BC024135.1 | - | 9 - 10 | 1104 | 76 | ||
| BC051055.1 | - | 9 - 10 | 1119 | 76 | ||
| BC057914.1 | - | 6 - 7 | 867 | 76 | ||
| BC088736.1 | - | 5 - 6 | 2452 | 76 | ||
| BC106183.1 | - | 9 - 10 | 1187 | 76 | ||
| BC132168.1 | - | 9 - 10 | 1203 | 40130445 | 76 | |
| BC132170.1 | - | 9 - 10 | 1203 | 40130447 | 76 | |
More Information
Set Membership: |
|

Coq6
Genomic Map