Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 2 - 3 | 471 | 104 | |
| GenBank mRNA | AK031041.1 | - | 2 - 3 | 474 | 104 | |
| AK142282.1 | - | 2 - 3 | 157 | 104 | ||
| AK146566.1 | - | 2 - 3 | 205 | 104 | ||
| AK164135.1 | - | 2 - 3 | 217 | 104 | ||
| BB620239.1 | - | 2 - 3 | 474 | 104 | ||
| BC019933.1 | - | 3 - 4 | 207 | 104 | ||
| BC047122.1 | - | 3 - 4 | 207 | 104 | ||
| BC052456.1 | - | 2 - 3 | 168 | 6830607 | 104 | |
| BC053718.1 | - | 2 - 3 | 187 | 104 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK164135.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map