Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 19 - 20 | 1919 | 105 | |
| GenBank mRNA | AK031041.1 | - | 19 - 20 | 1922 | 105 | |
| AK142282.1 | - | 19 - 20 | 1605 | 105 | ||
| AK164135.1 | - | 20 - 21 | 1777 | 105 | ||
| AK169439.1 | - | 3 - 4 | 1341 | 105 | ||
| BC052456.1 | - | 19 - 20 | 1616 | 6830607 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fcho2
Genomic Map