Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172598.3 | NP_766186.2 | 3 - 4 | 598 | 110 | |
| GenBank mRNA | AK036390.1 | - | 3 - 4 | 557 | 110 | |
| AK042443.1 | - | 2 - 3 | 2211 | 110 | ||
| AK052690.1 | - | 3 - 4 | 574 | 110 | ||
| BC063740.1 | - | 3 - 4 | 531 | 6328626 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdhd1
Genomic Map