Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172598.3 | NP_766186.2 | 12 - 13 | 1654 | 64 | |
| GenBank mRNA | AK036390.1 | - | 12 - 13 | 1724 | 64 | |
| AK052690.1 | - | 12 - 13 | 1741 | 64 | ||
| BC063740.1 | - | 12 - 13 | 1587 | 6328626 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdhd1
Genomic Map