Assay Details
| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172604.3 | NP_766192.1 | 4 - 5 | 570 | 99 | |
| GenBank mRNA | AK048423.1 | - | 4 - 5 | 571 | 99 | |
| AK054188.1 | - | 4 - 5 | 568 | 99 | ||
| BC030851.1 | - | 1 - 2 | 116 | 99 | ||
| BC051636.1 | - | 4 - 5 | 343 | 6508271 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scara3
Genomic Map