Assay Details
| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172605.3 | NP_766193.3 | 12 - 13 | 2339 | 70 | |
| GenBank mRNA | AK033390.1 | - | 12 - 13 | 2284 | 70 | |
| BE983697.1 | - | 2 - 3 | 70 | |||
More Information
Set Membership: |
|

Tdrd3
Genomic Map