| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500006563.mm.1 |
NM_001205353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006563.mm.1 |
NM_001205353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006563.mm.1 |
NM_172611.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006563.mm.1 |
NM_172611.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006563.mm.1 |
TC1500000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500006562.mm.1 |
NM_001205353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006562.mm.1 |
NM_001205353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006562.mm.1 |
NM_172611.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006562.mm.1 |
NM_172611.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006562.mm.1 |
TC1500000829.mm.1 |
MTA TC |