Assay Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172617.3 | NP_766205.1 | 7 - 8 | 878 | 55 | |
| GenBank mRNA | AK028222.1 | - | 7 - 8 | 879 | 55 | |
| BC025615.1 | - | 4 - 5 | 443 | 55 | ||
| BC058346.1 | - | 5 - 6 | 610 | 6832233 | 55 | |
| BC060611.1 | - | 6 - 7 | 842 | 6416823 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp523
Genomic Map