Assay Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172617.3 | NP_766205.1 | 8 - 9 | 949 | 84 | |
| GenBank mRNA | AK028222.1 | - | 8 - 9 | 950 | 84 | |
| BC025615.1 | - | 5 - 6 | 514 | 84 | ||
| BC058346.1 | - | 6 - 7 | 681 | 6832233 | 84 | |
| BC060611.1 | - | 7 - 8 | 913 | 6416823 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp523
Genomic Map