Assay Details
| Gene Symbol: Trerf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097623.1 | NP_001091092.1 | 14 - 15 | 3371 | 109 | |
| NM_172622.2 | NP_766210.1 | 11 - 12 | 3148 | 109 | ||
| GenBank mRNA | AK046822.1 | - | 9 - 10 | 2789 | 109 | |
| AY081774.1 | - | 10 - 11 | 3058 | 109 | ||
| BC059215.1 | - | 14 - 15 | 3371 | 6412734 | 109 | |
| BC108416.1 | - | 9 - 10 | 2831 | 5697521 | 109 | |
More Information
Set Membership: |
|

Trerf1
Genomic Map