Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172635.3 | NP_766223.2 | 5 - 6 | 783 | 87 | |
| GenBank mRNA | AK085013.1 | - | 5 - 6 | 725 | 87 | |
| AK087559.1 | - | 5 - 6 | 747 | 87 | ||
| AK150301.1 | - | 5 - 6 | 784 | 87 | ||
| AK154873.1 | - | 5 - 6 | 735 | 87 | ||
| AK170408.1 | - | 5 - 6 | 747 | 87 | ||
| AK170745.1 | - | 5 - 6 | 748 | 87 | ||
| AK170918.1 | - | 5 - 6 | 784 | 87 | ||
| BC058941.1 | - | 5 - 6 | 724 | 6825758 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Patl1
Genomic Map