Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172635.3 | NP_766223.2 | 7 - 8 | 974 | 62 | |
| GenBank mRNA | AK085013.1 | - | 7 - 8 | 916 | 62 | |
| AK087559.1 | - | 7 - 8 | 938 | 62 | ||
| AK150301.1 | - | 7 - 8 | 975 | 62 | ||
| AK154102.1 | - | 7 - 8 | 972 | 62 | ||
| AK170408.1 | - | 7 - 8 | 938 | 62 | ||
| AK170745.1 | - | 7 - 8 | 939 | 62 | ||
| AK170918.1 | - | 7 - 8 | 975 | 62 | ||
| BC047458.1 | - | 7 - 8 | 776 | 62 | ||
| BC058941.1 | - | 7 - 8 | 915 | 6825758 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Patl1
Genomic Map