Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172635.3 | NP_766223.2 | 15 - 16 | 2063 | 83 | |
| GenBank mRNA | AK085013.1 | - | 15 - 16 | 2005 | 83 | |
| AK087559.1 | - | 15 - 16 | 2027 | 83 | ||
| AK150301.1 | - | 15 - 16 | 2064 | 83 | ||
| AK154102.1 | - | 15 - 16 | 2061 | 83 | ||
| AK154873.1 | - | 15 - 16 | 2015 | 83 | ||
| AK161738.1 | - | 9 - 10 | 1025 | 83 | ||
| AK170408.1 | - | 15 - 16 | 2027 | 83 | ||
| AK170745.1 | - | 15 - 16 | 2028 | 83 | ||
| AK170918.1 | - | 15 - 16 | 2064 | 83 | ||
| BC047458.1 | - | 15 - 16 | 1865 | 83 | ||
| BC058941.1 | - | 15 - 16 | 2004 | 6825758 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Patl1
Genomic Map