Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081225.1 | NP_001074694.1 | 9 - 10 | 2597 | 77 | |
| GenBank mRNA | AK028324.1 | - | 5 - 6 | 548 | 77 | |
| AK030951.1 | - | 5 - 6 | 913 | 77 | ||
| AK031688.1 | - | 9 - 10 | 2569 | 77 | ||
| AK032405.1 | - | 5 - 6 | 552 | 77 | ||
| AK036700.1 | - | 7 - 8 | 2174 | 77 | ||
| AK147588.1 | - | 9 - 10 | 3070 | 77 | ||
| AK161809.1 | - | 5 - 6 | 960 | 77 | ||
| BC060679.1 | - | 5 - 6 | 514 | 77 | ||
| BC150999.1 | - | 9 - 10 | 2450 | 9087912 | 77 | |
| BC157937.1 | - | 9 - 10 | 2450 | 9088117 | 77 | |
| BC172097.1 | - | 9 - 10 | 2450 | 9054383 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam178a
Genomic Map