Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081225.1 | NP_001074694.1 | 12 - 13 | 2920 | 85 | |
| GenBank mRNA | AK028324.1 | - | 8 - 9 | 871 | 85 | |
| AK030951.1 | - | 8 - 9 | 1236 | 85 | ||
| AK031688.1 | - | 12 - 13 | 2892 | 85 | ||
| AK032405.1 | - | 8 - 9 | 875 | 85 | ||
| AK036700.1 | - | 10 - 11 | 2497 | 85 | ||
| AK147588.1 | - | 12 - 13 | 3393 | 85 | ||
| AK161809.1 | - | 8 - 9 | 1283 | 85 | ||
| BC060679.1 | - | 8 - 9 | 837 | 85 | ||
| BC150999.1 | - | 12 - 13 | 2773 | 9087912 | 85 | |
| BC157937.1 | - | 12 - 13 | 2773 | 9088117 | 85 | |
| BC172097.1 | - | 12 - 13 | 2773 | 9054383 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam178a
Genomic Map