Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 3 - 4 | 474 | 109 | |
| GenBank mRNA | AB057666.1 | - | 3 - 4 | 241 | 109 | |
| AB057667.1 | - | 3 - 4 | 241 | 109 | ||
| AK036678.1 | - | 3 - 4 | 469 | 109 | ||
| AK046790.1 | - | 447 | 109 | |||
| AK080096.1 | - | 3 - 4 | 438 | 109 | ||
| AK086974.1 | - | 3 - 4 | 405 | 109 | ||
| AK140272.1 | - | 3 - 4 | 524 | 109 | ||
| AK146079.1 | - | 3 - 4 | 287 | 109 | ||
| BC037003.1 | - | 3 - 4 | 383 | 109 | ||
| BC060956.1 | - | 3 - 4 | 331 | 30316511 | 109 | |
| CJ060534.1 | - | 3 - 4 | 524 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map