Assay Details
| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172662.3 | NP_766250.2 | 7 - 8 | 1136 | 72 | |
| GenBank mRNA | AK054272.1 | - | 7 - 8 | 1137 | 72 | |
| BC052069.1 | - | 4 - 5 | 701 | 5715781 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtdc1
Genomic Map