Assay Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172663.4 | NP_766251.3 | 9 - 10 | 1700 | 99 | |
| GenBank mRNA | AK047947.1 | - | 9 - 10 | 1368 | 99 | |
| AK141073.1 | - | 991 | 99 | |||
| AK164886.1 | - | 5 - 6 | 727 | 99 | ||
| BC048785.1 | - | 8 - 9 | 1139 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epc2
Genomic Map