Assay Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172663.4 | NP_766251.3 | 10 - 11 | 2040 | 73 | |
| GenBank mRNA | AK031025.1 | - | 10 - 11 | 2013 | 73 | |
| AK047947.1 | - | 10 - 11 | 1708 | 73 | ||
| AK141073.1 | - | 1331 | 73 | |||
| AK164886.1 | - | 6 - 7 | 1067 | 73 | ||
| BC048785.1 | - | 9 - 10 | 1479 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epc2
Genomic Map