Assay Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172663.4 | NP_766251.3 | 12 - 13 | 2340 | 93 | |
| GenBank mRNA | AK031025.1 | - | 12 - 13 | 2313 | 93 | |
| AK047947.1 | - | 12 - 13 | 2008 | 93 | ||
| AK141073.1 | - | 1631 | 93 | |||
| AK164886.1 | - | 8 - 9 | 1367 | 93 | ||
| BC048785.1 | - | 11 - 12 | 1779 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epc2
Genomic Map