Assay Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172673.3 | NP_766261.2 | 10 - 11 | 1115 | 72 | |
| GenBank mRNA | AK030883.1 | - | 10 - 11 | 871 | 72 | |
| AK044270.1 | - | 10 - 11 | 866 | 72 | ||
| AK146383.1 | - | 9 - 10 | 995 | 72 | ||
| BC038623.1 | - | 10 - 11 | 852 | 72 | ||
| BC057549.1 | - | 7 - 8 | 644 | 6416914 | 72 | |
| BC062889.1 | - | 10 - 11 | 1072 | 5698263 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Frmd5
Genomic Map