Assay Details
| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027375.1 | 2 - 3 | 366 | 73 | ||
| GenBank mRNA | AK039176.1 | - | 2 - 3 | 313 | 73 | |
| AK053736.1 | - | 2 - 3 | 285 | 73 | ||
| BC057158.1 | - | 2 - 3 | 277 | 5348717 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK039176.1,AK053736.1,BC057158.1 transcripts.
Set Membership: |
|

Ythdf3
Genomic Map