Assay Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172678.3 | NP_766266.3 | 1 - 2 | 265 | 67 | |
| GenBank mRNA | AK028571.1 | - | 1 - 2 | 193 | 67 | |
| AK031820.1 | - | 1 - 2 | 197 | 67 | ||
| AK075984.1 | - | 1 - 2 | 184 | 67 | ||
| AK145423.1 | - | 1 - 2 | 173 | 67 | ||
More Information
Set Membership: |
|

Acad9
Genomic Map